Learn More
Medchemexpress LLC Histamine | 51-45-6 | MFCD00005210 | ≥98.0% | 111.15 g/mol | C5H9N3 | 10 MG
SDP
Catalog No. 5000297636
Encompass_Preferred
Histamine analytical standard supplied as a powdered reference material for research and analytical applications. It provides a defined source of histamine for calibration, method development, and quality-control assays.
  • Analytical reference standard for histamine quantification.
  • High purity suitable for LC/GC/MS and HPLC assays.
  • Powder form with high aqueous solubility for easy reconstitution.
  • Sold in small laboratory pack sizes for analytical workflows.
  • Store at -20°C, protect from moisture and oxygen.
Catalog No. 50-002-97636 Supplier Medchemexpress LLC Supplier No. HYB1204R10MG
Histamine analytical standard supplied as a powdered reference material for research and analytical applications. It provides a defined source of histamine for calibration, method development, and quality-control assays.
  • Analytical reference standard for histamine quantification.
  • High purity suitable for LC/GC/MS and HPLC assays.
  • Powder form with high aqueous solubility for easy reconstitution.
  • Sold in small laboratory pack sizes for analytical workflows.
  • Store at -20°C, protect from moisture and oxygen.