Learn More
Medchemexpress LLC Histamine | 51-45-6 | MFCD00005210 | 99.99% | 25 MG
SDP
Catalog No. 5000414271
Encompass_Preferred
Histamine (Standard) is the analytical standard of Histamine (HY-B1204) intended for research and analytical applications. It is an agonist for histamine receptor and a vasodilator, an organic nitrogen compound involved in local immune responses, regulation of intestinal physiological functions, and acting as a neurotransmitter. Histamine affects the p38 MAPK/Akt signaling pathway, and exhibits antitumor, antioxidant, and anti-inflammatory activities.
  • Agonist for histamine receptor
  • Vasodilator
  • Involved in local immune responses
  • Regulates intestinal physiological functions
  • Acts as a neurotransmitter
  • Affects p38 MAPK/Akt signaling pathway
  • Exhibits antitumor, antioxidant, and anti-inflammatory activities
  • Can be used in research of acute myeloid leukemia
  • Can be used in research of malignant melanoma
  • Can be used in research of renal cell carcinoma
Catalog No. 50-004-14271 Supplier Medchemexpress LLC Supplier No. 25MGHYB1204R
Histamine (Standard) is the analytical standard of Histamine (HY-B1204) intended for research and analytical applications. It is an agonist for histamine receptor and a vasodilator, an organic nitrogen compound involved in local immune responses, regulation of intestinal physiological functions, and acting as a neurotransmitter. Histamine affects the p38 MAPK/Akt signaling pathway, and exhibits antitumor, antioxidant, and anti-inflammatory activities.
  • Agonist for histamine receptor
  • Vasodilator
  • Involved in local immune responses
  • Regulates intestinal physiological functions
  • Acts as a neurotransmitter
  • Affects p38 MAPK/Akt signaling pathway
  • Exhibits antitumor, antioxidant, and anti-inflammatory activities
  • Can be used in research of acute myeloid leukemia
  • Can be used in research of malignant melanoma
  • Can be used in research of renal cell carcinoma