Learn More
Biomatik Corporation Histamine Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50407
Encompass_Preferred
Histamine Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Histamine. The Histamine Polyclonal Antibody has been validated for the following applications: ELISA, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Please Inquire. Applications: ELISA, IHC. Target Synonyms: Histamine. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9221