Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Histamine bisphosphate monohydrate

Catalog No. 533105G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-310-5G 5 g
53-310-25G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-310-5G Supplier Sigma Aldrich Supplier No. 533105G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Histamine bisphosphate monohydrate

Specifications

CAS 51-74-1
Melting Point 128°C to 132°C
Molecular Formula C5H9N3 -+ 2H3PO4 -+ H2O
Linear Formula C5H9N3 -+ 2H3PO4 -+ H2O
MDL Number MFCD00150816
Quantity 5 g
Synonym 2-(4-Imidazolyl)ethylamine diphosphate salt; 2-(4-Imidazolyl)ethylamine diphosphate salt monohydrate; Histamine diphosphate salt monohydrate
Molecular Weight (g/mol) 325.15
Percent Purity ≥99.0% (NT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.