Learn More
Enzo Life Sciences Histamine Controls (1 Set)
SDP
Catalog No. NC1442281
Encompass_Preferred
Histamine Controls for use with Histamine ELISA Kit. Contents: 1 high, 1 medium, and 1 low control. Application: ELISA. Long Term Storage: -80°C
Catalog No. NC1442281 Supplier Enzo Life Sciences Supplier No. ENZKIT140CTL
Histamine Controls for use with Histamine ELISA Kit. Contents: 1 high, 1 medium, and 1 low control. Application: ELISA. Long Term Storage: -80°C