Learn More
Sigma Aldrich Fine Chemicals Biosciences Histamine dihydrochloride, 56-92-8, MFCD00012703, 5 g
Catalog No. NC0988147
Encompass
Empirical Formula (Hill Notation): C5H9N3 · 2HCl, Molecular Weight: 184.07, ≥99% (TLC), powder, mp: 249-252 °C, color: white to off-white, storage temp: 2-8°C, Synonym: 2-(4-Imidazolyl)ethylamine dihydrochloride.
Catalog No. NC0988147 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H72505G
Empirical Formula (Hill Notation): C5H9N3 · 2HCl, Molecular Weight: 184.07, ≥99% (TLC), powder, mp: 249-252 °C, color: white to off-white, storage temp: 2-8°C, Synonym: 2-(4-Imidazolyl)ethylamine dihydrochloride.