Learn More
Medchemexpress LLC Histamine dihydrochloride | 56-92-8 | 99.9% | 25 MG
SDP
Catalog No. 5000450669
Encompass_Preferred
Histamine dihydrochloride is an organic nitrogen compound that participates in local immune responses, regulates intestinal physiological functions, and acts as a neurotransmitter. It is also an agonist for the histamine receptor and a vasodilator. It affects the p38 MAPK/Akt signaling pathway and shows antitumor, antioxidant, and anti-inflammatory activities.
  • Regulates intestinal physiological functions
  • Acts as a neurotransmitter
  • Affects the p38 MAPK/Akt signaling pathway
  • Shows antitumor, antioxidant, and anti-inflammatory activities
  • Used in research for acute myeloid leukemia, malignant melanoma, and renal cell carcinoma
Catalog No. 50-004-50669 Supplier Medchemexpress LLC Supplier No. 25MGHYB0722R
Histamine dihydrochloride is an organic nitrogen compound that participates in local immune responses, regulates intestinal physiological functions, and acts as a neurotransmitter. It is also an agonist for the histamine receptor and a vasodilator. It affects the p38 MAPK/Akt signaling pathway and shows antitumor, antioxidant, and anti-inflammatory activities.
  • Regulates intestinal physiological functions
  • Acts as a neurotransmitter
  • Affects the p38 MAPK/Akt signaling pathway
  • Shows antitumor, antioxidant, and anti-inflammatory activities
  • Used in research for acute myeloid leukemia, malignant melanoma, and renal cell carcinoma