Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Histamine dihydrochloride | 56-92-8 | MFCD00012703 | 100.0% | 184.07 g/mol | C5H11Cl2N3 | 10 MG
SDP

Catalog No. 5000296696
Encompass_Preferred

Histamine dihydrochloride is the dihydrochloride salt of histamine supplied as an analytical standard for research and analytical applications. It is provided as a crystalline solid for use in method development, calibration, and biochemical assays.

  • High purity (99.96%) suitable for analytical use.
  • Molecular weight 184.07 g/mol and formula C5H11Cl2N3.
  • Available in small-scale packaging ideal for standards and method validation.
  • Common applications include biochemical research, immunology, and pharmaceutical analysis.
  • Supplied with datasheet and certificate of analysis for traceability.

Catalog No. 50-002-96696 Supplier Medchemexpress LLC Supplier No. HYB0722R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Histamine dihydrochloride is the dihydrochloride salt of histamine supplied as an analytical standard for research and analytical applications. It is provided as a crystalline solid for use in method development, calibration, and biochemical assays.

  • High purity (99.96%) suitable for analytical use.
  • Molecular weight 184.07 g/mol and formula C5H11Cl2N3.
  • Available in small-scale packaging ideal for standards and method validation.
  • Common applications include biochemical research, immunology, and pharmaceutical analysis.
  • Supplied with datasheet and certificate of analysis for traceability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.