Learn More
Enzo Life Sciences Histamine . dihydrochloride (5g). CAS: 56-92-8
SDP
Supplier:  Enzo Life Sciences ALX550132G005
Encompass_Preferred
Histamine triggers the inflammatory response. Alternative name: 1H-Imidazole-4-ethanamine . 2HCl. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in water (50mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9486