Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine diphosphate, MP Biomedicals™

Catalog No. ICN10034305 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10034305 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10034305 Supplier MP Biomedicals, Inc Supplier No. 0210034305
Add to Cart
Add to Cart

Chemical Identifiers

CAS 51-74-1
Molecular Formula C5H15N3O8P2
Molecular Weight (g/mol) 307.14
MDL Number MFCD00064316
InChI Key ZHIBQGJKHVBLJJ-UHFFFAOYSA-N
Synonym histamine acid phosphate, 2-1h-imidazol-4-yl ethanamine bis phosphate, histamine biphosphate, histamine dihydrogen phosphate, histamine phosphate 1:2, histamine phosphate, 1h-imidazole-4-ethanamine, phosphate 1:2, histamine phosphate usp, histamine positive, 4-2-aminoethyl imidazole bis dihydrogen phosphate
PubChem CID 65513
ChEBI CHEBI:51193
IUPAC Name 2-(1H-imidazol-5-yl)ethan-1-amine; bis(phosphoric acid)
SMILES OP(O)(O)=O.OP(O)(O)=O.NCCC1=CN=CN1

Specifications

Color White
Quantity 5 g
Formula Weight 325.15
Physical Form Crystalline Powder
Chemical Name or Material Histamine diphosphate
EINECSNumber 200-118-4
storageNote1 Store desiccated
Recommended Storage Store at -0°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.