Learn More
Cayman Chemical Histamine EIA Kit, 96 wells
SDP
Supplier:  Cayman Chemical 58965196
Encompass
Cayman’s Histamine EIA is a derivitization-amplified competitive enzyme immunoassay which detects histamine within the range from 40 to 5,500 pg/ml. The assay can be used for the analysis of histamine in blood (plasma or serum) without extraction or purification. The use of other sample types may require further processing or purification of the sample.
Catalog No. NC9813041