Learn More
Oxford Biomedical Research Inc Histamine EIA Kit
SDP
Catalog No. NC1762476
Encompass
This heterocyclic primary amine is derived form the decarboxylation of histidine. Histamine serves as a mediator of inflammation and hypersensitivity (anphylaxis) responses. Once sysnthesized by the histidine carboxylase enzyme, histamine is typically stored in metachromatic granules of basophils and mast cells (granulocytes) for later dispersion. Histamine is involved in a cascade physiological effects that are influenced by typical immune response and environmental factors.
Catalog No. NC1762476 Supplier Oxford Biomedical Research Inc Supplier No. EA31
This heterocyclic primary amine is derived form the decarboxylation of histidine. Histamine serves as a mediator of inflammation and hypersensitivity (anphylaxis) responses. Once sysnthesized by the histidine carboxylase enzyme, histamine is typically stored in metachromatic granules of basophils and mast cells (granulocytes) for later dispersion. Histamine is involved in a cascade physiological effects that are influenced by typical immune response and environmental factors.