Learn More
Bertin Corp Histamine Elisa kit
SDP
Supplier:  Bertin Corp A05890
Encompass_Preferred
Histamine is a natural amine present in many animal and human tissues. It was considered to be the mediator of immediate allergy. Histamine plays a role in various physiological processes, such as control of gastric acid secretion, neurotransmission and modulation of inflammatory and immunological reactions. This competitive enzyme-immunoassay uses a prederivation (derivate reagent with a structurre close to the spacer arm used for the immunogen preparation) of histamine that allows to enhance the sensitivity of the assay
Catalog No. NC1861321