Learn More
Abcam Histamine ELISA Kit, 96TEST
Supplier:  Abcam AB28533396TEST
Encompass_Preferred
Histamine ELISA Kit is a Competitive ELISA kit for the measurement of Histamine in Cell culture supernatant, Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5495