Learn More
Abcam Histamine ELISA kit, 1 x 96 Tests
Supplier:  Abcam AB2139751X96TESTS
Encompass_Preferred
The Histamine ELISA kit (colorimetric) is a competitive ELISA designed to quantify mouse, rat and human histamine (HIS) with a sensitivity of 0.068 ng/mL. - Wide dynamic range – quantifies 0.098–25 ng/mL - Compatible with multiple sample types – works with serum, EDTA plasma, urine, and cell culture media - Fast assay time – complete results in 2 hours 30 minutes - Cited in over 20 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1609568