Learn More
Apexbio Technology LLC Histamine Phosphate 51-74-1 10mM (in 1mL H2O)
SDP
Catalog No. 5000094675
Encompass_Preferred
Histamine Phosphate (51-74-1) is a small-molecule agonist targeting histamine receptors specifically the H1 and H2 receptors It is designed to activate these receptors thereby regulating vascular function and cellular signaling pathways Histamine Phosphate exerts its biological activity primarily through receptor-mediated signaling leading to vasodilation in arteries and capillaries In in vitro assays Histamine Phosphate demonstrates receptor activation with reported IC50 values for H1 and H2 receptors typically ranging from nanomolar to micromolar concentrations in receptor binding studies Based on these pharmacological properties Histamine Phosphate holds research potential in investigating vascular permeability inflammation gastric acid secretion and in characterizing histamine receptor signaling mechanisms
Catalog No. 50-000-94675 Supplier Apexbio Technology LLC Supplier No. B17705.1
Histamine Phosphate (51-74-1) is a small-molecule agonist targeting histamine receptors specifically the H1 and H2 receptors It is designed to activate these receptors thereby regulating vascular function and cellular signaling pathways Histamine Phosphate exerts its biological activity primarily through receptor-mediated signaling leading to vasodilation in arteries and capillaries In in vitro assays Histamine Phosphate demonstrates receptor activation with reported IC50 values for H1 and H2 receptors typically ranging from nanomolar to micromolar concentrations in receptor binding studies Based on these pharmacological properties Histamine Phosphate holds research potential in investigating vascular permeability inflammation gastric acid secretion and in characterizing histamine receptor signaling mechanisms