Learn More
Selleck Chemical LLC Histamine Phosphate S4117-100mg
SDP
Catalog No. 503109114
Encompass_Preferred
Histamine(Histamine diphosphate) acts directly on the blood vessels to dilate arteries and capillaries mediated by both H 1- and H 2-receptors Histamine Phosphate can be used to induce animal models of Gastric and Intestinal Ulcers
Catalog No. 50-310-9114 Supplier Selleck Chemical LLC Supplier No. S4117100MG
Histamine(Histamine diphosphate) acts directly on the blood vessels to dilate arteries and capillaries mediated by both H 1- and H 2-receptors Histamine Phosphate can be used to induce animal models of Gastric and Intestinal Ulcers