Learn More
Selleck Chemical LLC Histamine Phosphate S4117-50mg
SDP
Catalog No. 503109033
Encompass_Preferred
Histamine(Histamine diphosphate) acts directly on the blood vessels to dilate arteries and capillaries mediated by both H 1- and H 2-receptors Histamine Phosphate can be used to induce animal models of Gastric and Intestinal Ulcers
Catalog No. 50-310-9033 Supplier Selleck Chemical LLC Supplier No. S411750MG
Histamine(Histamine diphosphate) acts directly on the blood vessels to dilate arteries and capillaries mediated by both H 1- and H 2-receptors Histamine Phosphate can be used to induce animal models of Gastric and Intestinal Ulcers