Learn More
Medchemexpress LLC Histamine (Standard) | 51-45-6 | 100 MG
SDP
Catalog No. 5000413632
Encompass_Preferred
Histamine (Standard) is an analytical standard for research and analytical applications. It functions as an agonist for histamine receptors and a vasodilator, playing roles in local immune responses, intestinal regulation, and neurotransmission. It influences the p38 MAPK/Akt signaling pathway, exhibiting antitumor, antioxidant, and anti-inflammatory activities, and is used in research for acute myeloid leukemia, malignant melanoma, and renal cell carcinoma.
  • Analytical standard for research
  • Agonist for histamine receptor
  • Vasodilator and neurotransmitter
  • Antitumor, antioxidant, anti-inflammatory activities
  • Used in cancer research
  • Soluble in H2O at ≥ 34 mg/mL
Catalog No. 50-004-13632 Supplier Medchemexpress LLC Supplier No. HYB1204R100MG
Histamine (Standard) is an analytical standard for research and analytical applications. It functions as an agonist for histamine receptors and a vasodilator, playing roles in local immune responses, intestinal regulation, and neurotransmission. It influences the p38 MAPK/Akt signaling pathway, exhibiting antitumor, antioxidant, and anti-inflammatory activities, and is used in research for acute myeloid leukemia, malignant melanoma, and renal cell carcinoma.
  • Analytical standard for research
  • Agonist for histamine receptor
  • Vasodilator and neurotransmitter
  • Antitumor, antioxidant, anti-inflammatory activities
  • Used in cancer research
  • Soluble in H2O at ≥ 34 mg/mL