Learn More
Enzo Life Sciences SCREEN-WELL Histaminergic ligand library (500 µl/well)
SDP
Catalog No. 502010199
Encompass_Preferred
The Histaminergic and Melatonin Ligands Plate contains 41 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Histaminergic and Melatonin Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .
Catalog No. 50-201-0199 Supplier Enzo Life Sciences Supplier No. BML28160500
The Histaminergic and Melatonin Ligands Plate contains 41 ligands including endogenous neurotransmitters, agonists, antagonists, and marketed drugs. The library is ideal for screening or identifying recombinant orphan G protein coupled receptors, target validation, secondary screening, assay development, and other pharmacological applications. The Histaminergic and Melatonin Ligands plate is 1 plate of the larger Neurotransmitter Library (BML-2810) which contains over 680 ligands. Long Term Storage: -80°C. Applications: HTS .