Learn More
Cayman Chemical HIstatIn 3trIfluoroacetAT 10mg
SDP
Catalog No. 502915001
Encompass_Preferred
An antimicrobial peptide; active against C. albicans (LC50 = 9.2 µM); induces actin polymerization in a cell-free assay at 12 µM
Catalog No. 50-291-5001 Supplier Cayman Chemical Supplier No. 3698310 MG
An antimicrobial peptide; active against C. albicans (LC50 = 9.2 µM); induces actin polymerization in a cell-free assay at 12 µM