Learn More
CPC Scientific H-Asp-Ser-His-Ala-Lys-Arg-His-His-Gly-Tyr-Lys-Arg-Lys-Phe-His-Glu-Lys-His-His-Ser-His-Arg-Gly-Tyr-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific ATBP003A
Encompass
Salivary polypeptide shown to inhibit clostripain and a trypsin-like protease produced by Bacteroides gingivalis. Other proteases, however, are not inhibited by histatin-5. This peptide is thereby suggested to be a preventative against periodontal disease. ONE-LETTER SEQUENCE: DSHAKRHHGYKRKFHEKHHSHRGYMOLECULAR FORMULA: C133H195N51O33MOLECULAR WEIGHT:3036.5STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [104339-66-4], RESEARCH AREA: AntimicrobialREFERENCES: M.Nishikata et al., Biochem. Biophys. Res. Commun., 174, 625 (1991)
Catalog No. 50-210-8795