Learn More
Medchemexpress LLC HY-P0273A 1mg Medchemexpress, Histatin 5 (TFA) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0273A1MG
Encompass_Preferred
Medchemexpress, HY-P0273A 1mg Histatin 5 (TFA) CAS: Histatin 5 TFA inhibits the activity of the host matrix metalloproteinases MMP-2 and MMP-9 with IC 50 s of 0.57 and 0.25 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4090