Learn More
Cayman Chemical HIstatIn 5trIfluoroacetAT 10mg
SDP
Supplier:  Cayman Chemical 3672110 MG
Encompass_Preferred
An antimicrobial peptide; active against C. albicans (LC50 = 1.8 µM), as well as A. baumannii, P. aeruginosa, and E. cloacae (MICs = 38, 47, and 90 µM, respectively); topical application decreases fungal burden in a mouse model of C. albicans-induced vulvovaginal candidiasis at 20 µM
Catalog No. 50-290-9660