Learn More
Cayman Chemical Histatin 8 (trifluoroacetate salt), Purity: ≥98%, 10 mg
SDP
Catalog No. 502914889
Encompass_Preferred
An antimicrobial peptide; active against colistin-sensitive and -resistant strains of A. baumannii at 32 µg/ml; active against C. albicans and C. tropicalis (MICs = 1 and 5 mg/ml, respectively)
Catalog No. 50-291-4889 Supplier Cayman Chemical Supplier No. 3689110 MG
An antimicrobial peptide; active against colistin-sensitive and -resistant strains of A. baumannii at 32 µg/ml; active against C. albicans and C. tropicalis (MICs = 1 and 5 mg/ml, respectively)