Learn More
Bioworld Histidine Buffer 0.1M, pH 6.0 | 500mL
Catalog No. NC1742287
Encompass_Preferred
Histidine buffer has a concentration of 0.1M and a pH of 6.0. Histidine has a pKa of 6.2 but this can range from 5 - 8 when the amino acid is bound to other amino acids. Histidine makes a good amino acid buffer under physiological conditions, not under very acidic or very basic conditions, and is the best amino acid buffer within its buffering range.
Catalog No. NC1742287 Supplier Bioworld Supplier No. 401250352
Histidine buffer has a concentration of 0.1M and a pH of 6.0. Histidine has a pKa of 6.2 but this can range from 5 - 8 when the amino acid is bound to other amino acids. Histidine makes a good amino acid buffer under physiological conditions, not under very acidic or very basic conditions, and is the best amino acid buffer within its buffering range.