Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boston Bioproducts Inc 0.2 M Histidine Buffer, pH 7.0, 500 ml, Amino acid buffer, 0.22-micron filtered

Supplier:  Boston Bioproducts Inc BB2531500ML

Encompass

This 0.2 M Histidine Buffer, precisely adjusted to pH 7.0, is a high-purity, ready-to-use amino acid-based solution. Engineered for scientific applications, it effectively maintains stable pH conditions within a range of 5.0 to 8.0, making it ideal for critical procedures such as protein purification, enzyme assays, cell culture, and molecular biology techniques. The buffer is 0.22-micron filtered, ensuring reliability and reproducible results in sensitive biological research and development processes.

  • Buffer concentration: 0.2 M
  • Initial pH: 7.0
  • Volume: 500 ml
  • Buffer type: Amino acid
  • Effective buffering range: pH 5.0-8.0
  • Filtration: 0.22-micron
  • Storage temperature: 2-8°C
  • Quality control tests: Appearance, pH
  • Format: Ready-to-use solution
  • Primary applications: Protein purification, enzyme assays, cell culture, bioprocessing, molecular biology, drug development

Catalog No. NC2025362


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.