Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Histidyl-proline hydrochloride | 2415727-78-3 | 99.8% | 288.73 g·mol⁻¹ | C11H17ClN4O3 | 5 MG
SDP

Catalog No. 5000264742
Encompass_Preferred

His-Pro hydrochloride (histidyl-proline hydrochloride) is a research-grade dipeptide composed of histidyl and proline supplied as the hydrochloride salt. Offered as a white to off-white powder, it is intended for analytical and biochemical applications including peptide research and assay development.

  • High purity (~99.8%) suitable for analytical work.
  • Molecular weight 288.73 g·mol⁻¹.
  • Chemical formula C11H17ClN4O3.
  • Available in small, milligram-scale quantities for research use.
  • Solid powder form with recommended refrigerated storage to preserve stability.

Catalog No. 50-002-64742 Supplier Medchemexpress LLC Supplier No. HY111659A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

His-Pro hydrochloride (histidyl-proline hydrochloride) is a research-grade dipeptide composed of histidyl and proline supplied as the hydrochloride salt. Offered as a white to off-white powder, it is intended for analytical and biochemical applications including peptide research and assay development.

  • High purity (~99.8%) suitable for analytical work.
  • Molecular weight 288.73 g·mol⁻¹.
  • Chemical formula C11H17ClN4O3.
  • Available in small, milligram-scale quantities for research use.
  • Solid powder form with recommended refrigerated storage to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.