Learn More
BPS Bioscience Inc Histone H4 Peptide (1-21), 160 nmolePurity: ≥95% by HPLC
SDP
Catalog No. NC2605842
Encompass
A peptide substrate for Histone 4 R3 methyltransferases. The peptide corresponds to amino acid residues 1-21 of histone H4.
Catalog No. NC2605842 Supplier BPS Bioscience Inc Supplier No. 52014
A peptide substrate for Histone 4 R3 methyltransferases. The peptide corresponds to amino acid residues 1-21 of histone H4.