Learn More
Medchemexpress LLC Histo-blood group AB 5ug
SDP
Catalog No. 5000179300
Encompass_Preferred
The Histo-blood group ABO/ABO Protein is an enzyme encoded by the human ABO gene with glycosyltransferase activity ABO Protein is associated with a variety of infectious and non-communicable diseases ABO blood group can be used as a tumor marker Histo-blood group ABO/ABO Protein Human (HEK293 Fc) is the recombinant human-derived Histo-blood group ABO/ABO protein expressed by HEK293 with N-hFc labeled tag
Catalog No. 50-001-79300 Supplier Medchemexpress LLC Supplier No. HYP739265UG
The Histo-blood group ABO/ABO Protein is an enzyme encoded by the human ABO gene with glycosyltransferase activity ABO Protein is associated with a variety of infectious and non-communicable diseases ABO blood group can be used as a tumor marker Histo-blood group ABO/ABO Protein Human (HEK293 Fc) is the recombinant human-derived Histo-blood group ABO/ABO protein expressed by HEK293 with N-hFc labeled tag