Learn More
Electron Microscopy Sciences Histo-Clear, 1Gal
SDP
Catalog No. 5032950
Encompass_Preferred
Histo-Clear is distilled essential oils that is food grade. t is an excellent non-toxic clearing agent that is replacing Xylene.
Catalog No. 50-329-50 Supplier Electron Microscopy Sciences Supplier No. 6411004
Histo-Clear is distilled essential oils that is food grade. t is an excellent non-toxic clearing agent that is replacing Xylene.