Learn More
National Diagnostics HISTO-CLEAR 5 GLHISTO-CLEAR
SDP
Catalog No. 5089990149
Encompass
Histo-Clear, Orange oil based clearing agent providing rapid clearing without the toxic hazard of xylene, 5 GL
Catalog No. 50-899-90149 Supplier National Diagnostics Supplier No. HS2005GLL
Histo-Clear, Orange oil based clearing agent providing rapid clearing without the toxic hazard of xylene, 5 GL