Learn More
National Diagnostics HISTO-CLEAR II 1 GL
SDP
Catalog No. 5089990150
Encompass
Histo-Clear II, Xylene substitute composed of ultra-refined light paraffin oil and orange oil. Greatly reduced citrus odor, 1 GL
Catalog No. 50-899-90150 Supplier National Diagnostics Supplier No. HS2021GLL
Histo-Clear II, Xylene substitute composed of ultra-refined light paraffin oil and orange oil. Greatly reduced citrus odor, 1 GL