Learn More
Electron Microscopy Sciences Histo-Clear II, 1Gal
SDP
Catalog No. 5032948
Encompass_Preferred
Histo-Clear II is a mixture of Aliphatic Hydrocarbon and Distilled essential oils. It is an excellent non-toxic clearing agent that is replacing Xylene.
Catalog No. 50-329-48 Supplier Electron Microscopy Sciences Supplier No. 6411104
Histo-Clear II is a mixture of Aliphatic Hydrocarbon and Distilled essential oils. It is an excellent non-toxic clearing agent that is replacing Xylene.