Learn More
Electron Microscopy Sciences Histo-Clear II, 1L
SDP
Catalog No. 5032947
Encompass_Preferred
Histo-Clear II is a mixture of Aliphatic Hydrocarbon and Distilled essential oils that is food grade with a reduced citrus odor. It is an excellent non-toxic clearing agent that is replacing Xylene.
Catalog No. 50-329-47 Supplier Electron Microscopy Sciences Supplier No. 6411101
Histo-Clear II is a mixture of Aliphatic Hydrocarbon and Distilled essential oils that is food grade with a reduced citrus odor. It is an excellent non-toxic clearing agent that is replacing Xylene.