Learn More
Electron Microscopy Sciences Histo-Clear II Non-toxic Clearing Agent 5 GL
SDP
Catalog No. 5032952
Encompass_Preferred
Histo-Clear (Distilled essential oils –food grade) and Histo-Clear II (A mixtureof Aliphatic Hydrocarbon and Distilled essential oils – food grade, reduced citrus odor) are excellent non-toxic clearing agents replacing Xylene.
Catalog No. 50-329-52 Supplier Electron Microscopy Sciences Supplier No. 6411110
Histo-Clear (Distilled essential oils –food grade) and Histo-Clear II (A mixtureof Aliphatic Hydrocarbon and Distilled essential oils – food grade, reduced citrus odor) are excellent non-toxic clearing agents replacing Xylene.