Learn More
Electron Microscopy Sciences Histo-Container Opaque, 500 ml, 110 mm height, 88 mm I.D.
SDP
Catalog No. 5026550
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 500 ml
Height: 110 mm
I.D.: 88 mm
Catalog No. 50-265-50 Supplier Electron Microscopy Sciences Supplier No. 6423320
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 500 ml
Height: 110 mm
I.D.: 88 mm