Learn More
Electron Microscopy Sciences Histo-Container Opaque, 8L, 245 mm height, 230 mm I.D.
SDP
Catalog No. 5026557
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with snap-top lids and bucket carry handles and without graduations. Made of polyethylene.
Capacity: 8L
Height: 245 mm
I.D.: 230 mm
Catalog No. 50-265-57 Supplier Electron Microscopy Sciences Supplier No. 6423328
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with snap-top lids and bucket carry handles and without graduations. Made of polyethylene.
Capacity: 8L
Height: 245 mm
I.D.: 230 mm