Learn More
Electron Microscopy Sciences Histo-Container Opaque, 1L, 135 mm height, 110 mm I.D.
SDP
Catalog No. 5026549
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 1L
Height: 135 mm
I.D.: 110 mm
Catalog No. 50-265-49 Supplier Electron Microscopy Sciences Supplier No. 6423322
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 1L
Height: 135 mm
I.D.: 110 mm