Learn More
Electron Microscopy Sciences Histo-Container Semiclear, 40 ml, 40 mm height, 42 mm I.D.
SDP
Catalog No. 5026555
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps.
Capacity: 40 ml
Height: 40 mm
I.D.: 42 mm
Catalog No. 50-265-55 Supplier Electron Microscopy Sciences Supplier No. 6423312
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps.
Capacity: 40 ml
Height: 40 mm
I.D.: 42 mm