Learn More
Electron Microscopy Sciences Histo-Container Semiclear, 90 ml, 75 mm height, 42 mm I.D.
SDP
Catalog No. 5026553
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 90 ml
Height: 75 mm
I.D.: 42 mm
Catalog No. 50-265-53 Supplier Electron Microscopy Sciences Supplier No. 6423316
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 90 ml
Height: 75 mm
I.D.: 42 mm