Learn More
Electron Microscopy Sciences Histo-Container Semiclear, 120 ml, 75 mm height, 48 mm I.D.
SDP
Catalog No. 5026552
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 120 ml
Height: 75 mm
I.D.: 48 mm
Catalog No. 50-265-52 Supplier Electron Microscopy Sciences Supplier No. 6423317
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with tight-fitting screw caps and graduated on three sides.
Capacity: 120 ml
Height: 75 mm
I.D.: 48 mm