Learn More
Electron Microscopy Sciences Histo-Container White, 2L, 120 mm height, 184 mm I.D.
SDP
Catalog No. 5026548
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with snap-top lids and bucket carry handles and without graduations.
Capacity: 2L
Height: 120 mm
I.D.: 184 mm
Catalog No. 50-265-48 Supplier Electron Microscopy Sciences Supplier No. 6423324
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with snap-top lids and bucket carry handles and without graduations.
Capacity: 2L
Height: 120 mm
I.D.: 184 mm