Learn More
Electron Microscopy Sciences Histo-Container White, 5 L, 190 mm height, 126 mm I.D.
SDP
Catalog No. 5026546
Encompass_Preferred
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with snap-top lids and bucket carry handles and without graduations.
Capacity: 5 L
Height: 190 mm
I.D.: 126 mm
Catalog No. 50-265-46 Supplier Electron Microscopy Sciences Supplier No. 6423326
For the storage and transport of histology specimens, these containers are made from durable polypropylene and have leak proof seal caps. Supplied with snap-top lids and bucket carry handles and without graduations.
Capacity: 5 L
Height: 190 mm
I.D.: 126 mm