Learn More
Lab Storage Systems Inc Histo Cool Block Cooling System, for 30 Blocks, Blue
SDP
Catalog No. NC0722159
Encompass
The Histo-Cool was designed to help keep ice frozen throughout the day thus minimizing the need for histology personnel having to go to the freezer and hope there are spare ice trays available. The double wall design allows Freezer gel in sides and bottom keep ice frozen longer than traditional chilling methods. When kept in freezer overnight, the Histo-Cool stays frozen 6-8 hours on benchtop.
Catalog No. NC0722159 Supplier Lab Storage Systems Inc Supplier No. HC30
The Histo-Cool was designed to help keep ice frozen throughout the day thus minimizing the need for histology personnel having to go to the freezer and hope there are spare ice trays available. The double wall design allows Freezer gel in sides and bottom keep ice frozen longer than traditional chilling methods. When kept in freezer overnight, the Histo-Cool stays frozen 6-8 hours on benchtop.