Learn More
Ted Pella Inc PELCO Histo/Cyto-freeze™, 10 oz.
SDP
Catalog No. NC0579748
Encompass
Excellent for cooling liquid, semi-solid and solid specimens. Ideal for ultra-rapid freezing of fresh tissue, cooling of liquids to induce precipitation prior to filtering and cooling of field samples. PELCO Histo/Cyto-Freeze™ delivers instant flash freezing for cryostat sectioning. It cools down to -60°F (-50°C) in just three seconds. To cut frozen sections, mount sample directly on the microtome holder, simply spray to flash freeze and section. Time saving procedure and prevents water crystals built up caused by slower methods. The provided nozzle delivers the freezing action exactly where it is needed. Contains 10 oz. pure, 100% virgin tetrafluoroethane for enhanced freezing and cooling performance.
Catalog No. NC0579748 Supplier Ted Pella Inc Supplier No. 27219
Excellent for cooling liquid, semi-solid and solid specimens. Ideal for ultra-rapid freezing of fresh tissue, cooling of liquids to induce precipitation prior to filtering and cooling of field samples. PELCO Histo/Cyto-Freeze™ delivers instant flash freezing for cryostat sectioning. It cools down to -60°F (-50°C) in just three seconds. To cut frozen sections, mount sample directly on the microtome holder, simply spray to flash freeze and section. Time saving procedure and prevents water crystals built up caused by slower methods. The provided nozzle delivers the freezing action exactly where it is needed. Contains 10 oz. pure, 100% virgin tetrafluoroethane for enhanced freezing and cooling performance.