Learn More
Visikol Inc Visikol HISTO-M Starter Kit
Catalog No. 501616412
Encompass_Preferred
Laboratory Reagent, Histological Reagent, Tissue Clearing Reagent, Visikol HISTO-M Starter Kit. With the Visikol HISTO tissue clearing approach you are now able to rapidly and easily image tissues in 3D without damaging the underlying cellular morphology. The Visikol HISTO approach has been designed to pair with immunolabeling and is reversible so that 3D histology can be followed up with traditional 2D histology. The Visikol HISTO tissue clearing approach for 3D tissue cultures is comprised of one reagent called Visikol HISTO-M. In our Starter Kit we have combined 30 mL of Visikol HISTO-M with the four pre-treatment solutions required to process 3D tissue cultures for immunolabeling. All you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of micro-tissues.
Catalog No. 50-161-6412 Supplier Visikol Inc Supplier No. HMSK1
Laboratory Reagent, Histological Reagent, Tissue Clearing Reagent, Visikol HISTO-M Starter Kit. With the Visikol HISTO tissue clearing approach you are now able to rapidly and easily image tissues in 3D without damaging the underlying cellular morphology. The Visikol HISTO approach has been designed to pair with immunolabeling and is reversible so that 3D histology can be followed up with traditional 2D histology. The Visikol HISTO tissue clearing approach for 3D tissue cultures is comprised of one reagent called Visikol HISTO-M. In our Starter Kit we have combined 30 mL of Visikol HISTO-M with the four pre-treatment solutions required to process 3D tissue cultures for immunolabeling. All you need to do is add your immunolabels and nuclear stains. This kit comes with enough material to process three 96-well plates of micro-tissues.