Learn More
Electron Microscopy Sciences Histo-Mark Pen, fine line, 1 ea
SDP
Catalog No. NC0240789
Encompass_Preferred
The Histo-Mark pen is designed to work like a pen. It is a fine-line marker that can be used on any surface. The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene. Marking quality remains consistent throughout the life of the ink barrel.
Catalog No. NC0240789 Supplier Electron Microscopy Sciences Supplier No. 7215501
The Histo-Mark pen is designed to work like a pen. It is a fine-line marker that can be used on any surface. The Histo-Mark pen is solvent resistent, especially to alcohol, acetone, xylene, xylene subsite and toluene. Marking quality remains consistent throughout the life of the ink barrel.