Learn More
Electron Microscopy Sciences Histo-Mount 450 ML
SDP
Catalog No. 5032234
Encompass_Preferred
A mixture of xylene - mixed isomers and non hazardous ingredients to offer a very good synthetic mountant. Once dried the refractive index is 1.58.
Catalog No. 50-322-34 Supplier Electron Microscopy Sciences Supplier No. 1799505
A mixture of xylene - mixed isomers and non hazardous ingredients to offer a very good synthetic mountant. Once dried the refractive index is 1.58.