Learn More
Electron Microscopy Sciences Histo Orientator
SDP
Catalog No. 5026523
Encompass_Preferred
The convenient 2"x2" (51x51 mm) heated surface can be used for either "de-wrinkling" of paraffin sections or for the "rapid attachment" of fresh frozen sections. A quick touch of the slide to the hot plate will remove wrinkles and bubbles on your sections. You can also keep a unit on your cryostat to ensure solid attachment of frozen sections to the slide which prevents loosening of the tissue during agitation in hand staining. Temperature range: Ambient to 65 degree C.
Catalog No. 50-265-23 Supplier Electron Microscopy Sciences Supplier No. 63205
The convenient 2"x2" (51x51 mm) heated surface can be used for either "de-wrinkling" of paraffin sections or for the "rapid attachment" of fresh frozen sections. A quick touch of the slide to the hot plate will remove wrinkles and bubbles on your sections. You can also keep a unit on your cryostat to ensure solid attachment of frozen sections to the slide which prevents loosening of the tissue during agitation in hand staining. Temperature range: Ambient to 65 degree C.